Skip to Content
Merck
All Photos(1)

Key Documents

940364

Sigma-Aldrich

GPhos Pd G4 ChemBeads

new

Synonym(s):

Methanesulfonato[Dicyclohexyl[3-(1,1-dimethylethoxy)-6-methoxy-2′,6′-bis(1-methylethyl)[1,1′-biphenyl]-2-yl]phosphine](2′-methylamino-1,1′-biphenyl-2-yl)palladium(II) ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H68NO5PPdS
Molecular Weight:
920.53
UNSPSC Code:
12352100

form

solid

Quality Level

composition

, 4-6 wt. % (loading)

reaction suitability

reagent type: catalyst

SMILES string

COC(C(C1=C(C=CC=C1C(C)C)C(C)C)=C2[P](C3CCCCC3)([Pd]4(C5=C(C=CC=C5)C6=C(C=CC=C6)[NH]4C)OS(C)(=O)=O)C7CCCCC7)=CC=C2OC(C)(C)C

InChI

InChI=1S/C35H53O2P.C13H12N.CH4O3S.Pd/c1-24(2)28-20-15-21-29(25(3)4)32(28)33-30(36-8)22-23-31(37-35(5,6)7)34(33)38(26-16-11-9-12-17-26)27-18-13-10-14-19-27;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h15,20-27H,9-14,16-19H2,1-8H3;2-7,9-10,14H,1H3;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

LJIBRSQNADCZDI-UHFFFAOYSA-M

General description

The ChemBeads product of the GPhos ligand combined with the Buchwald Fourth Generation Palladacycle. The GPhos motif gives superb activity while having greater stability relative to other superbulky Buchwald ligands. Loaded at 5% wt. on glass beads for use in high-throughput expermentation (HTE).

Application

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

For general uses, product is also available in powdered form 920479

Features and Benefits

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott D McCann et al.
Journal of the American Chemical Society, 142(35), 15027-15037 (2020-08-14)
We have developed a new dialkylbiaryl monophosphine ligand, GPhos, that supports a palladium catalyst capable of promoting carbon-nitrogen cross-coupling reactions between a variety of primary amines and aryl halides; in many cases, these reactions can be carried out at room
Bryan T Ingoglia et al.
Tetrahedron, 75(32), 4199-4211 (2020-01-04)
Over the past three decades, Pd-catalyzed cross-coupling reactions have become a mainstay of organic synthesis. In particular, catalysts derived from biaryl monophosphines have shown wide utility in forming C-N bonds under mild reaction conditions. This work summarizes a variety of
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents
Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service