940119
Copper(I) Chloride ChemBeads
15 wt% loading
Synonym(s):
Copper monochloride ChemBeads, CuCl ChemBeads, Cuprous chloride ChemBeads
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CuCl
CAS Number:
Molecular Weight:
99.00
MDL number:
UNSPSC Code:
12352100
form
solid
Quality Level
composition
15 wt% loading
reaction suitability
reagent type: catalyst
SMILES string
[Cu+].[Cl-]
InChI key
OXBLHERUFWYNTN-UHFFFAOYSA-M
General description
Copper(I) chloride is a lewis acid and a useful source of copper(I) in chemical synthesis. Loaded on glass beads for use in high-throughput expermentation (HTE).
Application
Copper(I) chloride (Cuprous chloride, CuCl) has been used: In the synthesis of 7-((tert-butyldimethylsilyl)oxy)hepta-2,4-diyn-1-ol.
In the synthesis of CuInSe nanocrystals.
To catalyze the synthesis of benzothiazolethiones from iodoanilines, isocyanide and potassium sulfide.
Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
For general uses, product is also available in powdered form (212946)
In the synthesis of CuInSe nanocrystals.
To catalyze the synthesis of benzothiazolethiones from iodoanilines, isocyanide and potassium sulfide.
Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
For general uses, product is also available in powdered form (212946)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Beeraiah Baire et al.
Nature protocols, 8(3), 501-508 (2013-02-16)
The hexadehydro-Diels-Alder (HDDA) cascade enables the synthesis of complex benzenoid products with various substitution patterns through aryne intermediates. The first stage of this cascade involves the generation of a highly reactive ortho-benzyne intermediate by a net [4+2] cycloisomerization of a
Bryan T Ingoglia et al.
Tetrahedron, 75(32), 4199-4211 (2020-01-04)
Over the past three decades, Pd-catalyzed cross-coupling reactions have become a mainstay of organic synthesis. In particular, catalysts derived from biaryl monophosphines have shown wide utility in forming C-N bonds under mild reaction conditions. This work summarizes a variety of
Bonil Koo et al.
Journal of the American Chemical Society, 131(9), 3134-3135 (2009-02-17)
The synthesis of monodisperse chalcopyrite (tetragonal) CuInSe(2) nanocrystals is reported. The nanocrystals have trigonal pyramidal shape, and they exhibited a common crystallographic orientation when drop-cast onto carbon substrates. A crystallographic model for the nanocrystals was developed. The nanocrystals are bounded
Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service