Skip to Content
Merck
All Photos(1)

Key Documents

93770

Sigma-Aldrich

Tuerk solution

for microscopy (for counting of Leukozytes)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€28.20
1 L
€150.00

€28.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€28.20
1 L
€150.00

About This Item

MDL number:
UNSPSC Code:
12171500
NACRES:
MA.02

€28.20


Please contact Customer Service for Availability

Request a Bulk Order

grade

for microscopy (for counting of Leukozytes)

Quality Level

form

liquid

refractive index

n20/D 1.334

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service