Skip to Content
Merck
All Photos(1)

Key Documents

932175

Sigma-Aldrich

Potassium hexachloroplatinate(IV) CHEMBEADS

Synonym(s):

Dipotassium hexachloroplatinate, Dipotassium platinum hexachloride, Platinic potassium chloride, Platinum potassium chloride, Potassium chloroplatinate, Potassium platinum(IV) chloride, Potassium platinum(IV) hexachloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€125.00

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€125.00

About This Item

Empirical Formula (Hill Notation):
K2PtCl6
CAS Number:
Molecular Weight:
485.99
UNSPSC Code:
12352302
NACRES:
NA.22

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

Quality Level

storage temp.

2-8°C

SMILES string

[K][K].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2K.Pt/h6*1H;;;/q;;;;;;2*+1;+4/p-6

InChI key

DPAIVKJGTXERIM-UHFFFAOYSA-H

General description

Potassium hexachloroplatinate (K2PtCl6) is the inorganic compound that is commonly used as a platinum (Pt) precursor for the preparation of platinum-based compounds.

Application

Potassium hexachloroplatinate (K2PtCl6) can be used to synthesize:
  • K2PtCl6-PEG/SiO2 complex catalytic system for vapor-phase hydrosilylation reaction.
  • Pt-decorated nickel pyrophosphate ß-Ni2P2O7/Pt and nickel phosphate Ni3(PO4)2/Pt catalysts for the hydrogen evolution reaction (HER) by water splitting.
  • Polypyrrole-platinum complex sphere catalyst for the decomposition of hydrogen peroxide to form oxygen.
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

Product is also available as neat precatalyst (206067)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Versatile Methods to Dispense Submilligram Quantities of Solids Using Chemical-Coated Beads for High-Throughput Experimentation
Martin, et al.
Organic Process Research & Development, 2, 1900?1907-1900?1907 (2019)
Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service