Skip to Content
Merck
All Photos(1)

Key Documents

92588

Supelco

3,4-Dihydroxybenzaldehyde

analytical standard

Synonym(s):

Protocatechualdehyde

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€50.50

€50.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€50.50

About This Item

Linear Formula:
(HO)2C6H3CHO
CAS Number:
Molecular Weight:
138.12
Beilstein:
774381
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€50.50


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

150-155 °C
150-157 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Oc1ccc(C=O)cc1O

InChI

1S/C7H6O3/c8-4-5-1-2-6(9)7(10)3-5/h1-4,9-10H

InChI key

IBGBGRVKPALMCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong Rak Lee et al.
Archives of biochemistry and biophysics, 564, 203-210 (2014-12-03)
Protocatechualdehyde (PCA) is one of the important compounds found in barley, green cavendish bananas and grapevine leaves. PCA shows anti-cancer activities in breast, leukemia and colorectal cancer cells. Previous study reported that PCA exerts anti-cancer activity through down-regulating cyclin D1
Liang Zhang et al.
Toxicology and applied pharmacology, 283(1), 50-56 (2015-01-15)
An abnormal high mobility group box 1 (HMGB1) activation and a decrease in receptor for advanced glycation end-product (RAGE) play a key role in the pathogenesis of pulmonary fibrosis. Protocatechuic aldehyde (PA) is a naturally occurring compound, which is extracted
Aditya M Kunjapur et al.
Journal of the American Chemical Society, 136(33), 11644-11654 (2014-07-31)
Aromatic aldehydes are useful in numerous applications, especially as flavors, fragrances, and pharmaceutical precursors. However, microbial synthesis of aldehydes is hindered by rapid, endogenous, and redundant conversion of aldehydes to their corresponding alcohols. We report the construction of an Escherichia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service