Skip to Content
Merck
All Photos(1)

Key Documents

92587

Millipore

EC O157:H7 ChromoSelect Agar, Modified

NutriSelect® Plus, suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

28.85 G
€68.20
250 G
€361.00

€68.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
28.85 G
€68.20
250 G
€361.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

€68.20


Please contact Customer Service for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12 g/L
bile salts mixture, 1.5 g/L
chromogenic mixture, 0.25 g/L
sodium lauryl sulfate, 0.1 g/L
sorbitol, 7 g/L
tryptone, 5 g/L
yeast extract, 3 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

6.8±0.2 (25 °C)

application(s)

agriculture
food and beverages

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
coliforms

General description

A chromogenic medium recommended for isolation and differentiation of E. coli O157:H7 from food and environmental samples.

Application

for isolation and differentiation of E. coli O157:H7 from food and environmental samples

Preparation Note

Suspend 28.85 g in 1 liter distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C ) for 15 minutes. Cool to 50°C and pour into sterile petri plates. This medium can be made more selective by aseptically adding 0.25 ml of 1% potassium tellurite solution (Cat. No. 17774) to 1 liter molten and cooled medium (45-50°C).

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service