Skip to Content
Merck
All Photos(1)

Documents

92574

Supelco

Mefenamic acid

analytical standard

Synonym(s):

2-[(2,3-Dimethylphenyl)amino]benzoic acid, N-(2,3-Xylyl)anthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO2
CAS Number:
Molecular Weight:
241.29
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cc1cccc(Nc2ccccc2C(O)=O)c1C

InChI

1S/C15H15NO2/c1-10-6-5-9-13(11(10)2)16-14-8-4-3-7-12(14)15(17)18/h3-9,16H,1-2H3,(H,17,18)

InChI key

HYYBABOKPJLUIN-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
An NSAID. Circumvents MRP-mediated multidrug resistance. Specifically and significantly potentiates the cytotoxicity of anthracyclines as well as teniposide, VP-16 and vincristine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renata Colombo et al.
Journal of environmental management, 167, 206-213 (2015-12-22)
Mefenamic acid (MEF) is a non-steroidal anti-inflammatory drug indicated for relief of mild to moderate pain, and for the treatment of primary dysmenorrhea. The presence of MEF in raw and sewage waters has been detected worldwide at concentrations exceeding the
Umang V Shah et al.
International journal of pharmaceutics, 472(1-2), 140-147 (2014-06-15)
The role of surface properties, influenced by particle processing, in particle-particle interactions (powder cohesion) is investigated in this study. Wetting behaviour of mefenamic acid was found to be anisotropic by sessile drop contact angle measurements on macroscopic (>1cm) single crystals
Rachel Moll et al.
The Cochrane database of systematic reviews, (3)(3), CD007553-CD007553 (2011-03-18)
Mefenamic acid is a non-steroidal anti-inflammatory drug (NSAID). It is most often used for treating pain of dysmenorrhoea in the short term (seven days or less), as well as mild to moderate pain including headache, dental pain, postoperative and postpartum
C Reichert et al.
Clinical toxicology (Philadelphia, Pa.), 52(6), 629-634 (2014-05-23)
Seizures during intoxications with pharmaceuticals are a well-known complication. However, only a few studies report on drugs commonly involved and calculate the seizure potential of these drugs. To identify the pharmaceutical drugs most commonly associated with seizures after single-agent overdose
Harini Venkataraman et al.
Bioorganic & medicinal chemistry, 22(20), 5613-5620 (2014-07-08)
Cytochrome P450 BM3 mutants are promising biocatalysts for the production of drug metabolites. In the present study, the ability of cytochrome P450 BM3 mutants to produce oxidative metabolites of structurally related NSAIDs meclofenamic acid, mefenamic acid and tolfenamic acid was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service