Skip to Content
Merck
All Photos(1)

Key Documents

91298

Supelco

3-Hydroxyisovaleryl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-2-(3-hydroxy-3-methyl-1-oxobutoxy)-N,N,N-trimethyl-1-propanaminium inner salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€274.00

€274.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€274.00

About This Item

Empirical Formula (Hill Notation):
C12H23NO5
CAS Number:
Molecular Weight:
261.31
Beilstein:
10549761
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€274.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

optical activity

[α]/D -17.0±2.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

≤15% water (calc. on elemental analysis)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CC(C)(O)C)=O)CC([O-])=O

InChI

1S/C12H23NO5/c1-9(2)6-11(16)18-12(17,7-10(14)15)8-13(3,4)5/h9,17H,6-8H2,1-5H3

InChI key

DHRNMNHPSWBJEN-UHFFFAOYSA-N

Biochem/physiol Actions

3-Hydroxyisovaleryl-L-carnitine is an endogenous metabolite that can result from impairment in the leucine catabolic pathway. 3-Hydroxyisovaleryl-L-carnitine was observed in plasma and urine of patient with isolated 3-methylcrotonyl-CoA carboxylase deficiency and is used as biomarker of biotin deficiency in humans and other metabolic disorders.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urinary Excretion of 3-Hydroxyisovaleryl Carnitine Is an Early and Sensitive Indicator of Marginal Biotin Deficiency in Humans.
Stratton, S.L., et al.
The Journal of Nutrition, 141, 353-358 (2011)
3-Hydroxyisovalerylcarnitine in 3-methylcrotonyl-CoA carboxylase deficiency.
van Hove, J.L.K., et al.
Journal of inherited metabolic disease, 18, 592-601 (1995)
Plasma concentration of 3-hydroxyisovaleryl carnitine is an early and sensitive indicator of marginal biotin deficiency in humans.
Stratton, S.L., et al.
American Journal of Clinical Nutrition, 92, 1399-1405 (2010)
Quantitative Measurement of Urinary Excretion of 3-Hydroxyisovaleryl Carnitine by LC-MS/MS as an Indicator of Biotin Status in Humans.
Horvath, T.D., et al.
Analytical Chemistry, 82, 9543-9548 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service