Skip to Content
Merck
All Photos(1)

Key Documents

90969

Supelco

Ricinoleic acid

analytical standard

Synonym(s):

(R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€136.00

€136.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€136.00

About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH2CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
298.46
Beilstein:
1727813
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€136.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

optical activity

[α]/D 6.5±1.5°, c = 2 in acetone

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.469-1.473

density

0.940 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

functional group

carboxylic acid
hydroxyl

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCC[C@@H](O)C\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O3/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21/h9,12,17,19H,2-8,10-11,13-16H2,1H3,(H,20,21)/b12-9-/t17-/m1/s1

InChI key

WBHHMMIMDMUBKC-QJWNTBNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

435.2 °F - closed cup

Flash Point(C)

224 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Salvadora Ortega-Requena et al.
Journal of the science of food and agriculture, 94(11), 2308-2316 (2014-01-10)
Different strategies for the solvent-free enzymatic production of polyglycerol polyricinoleate (PGPR) were explored in an attempt to simplify and improve the process. Besides the conventional procedure (obtaining polyricinoleic acid, followed by its esterification with polyglycerol), two alternative methods are proposed:
Christina M Jones et al.
Journal of proteome research, 14(2), 917-927 (2015-01-09)
Ovarian cancer is a deadly disease killing more than any other gynecologic cancer. Nonspecific symptoms, combined with a lack of early detection methods, contribute to late diagnosis and low five-year survival rates. High-grade serous carcinoma (HGSC) is the most common
Sašo Gyergyek et al.
Acta chimica Slovenica, 61(3), 488-496 (2014-10-07)
Magnetic CoFe(2)O(4) nanoparticles were synthesized by co-precipitation at 80 °C. This co-precipitation was achieved by the rapid addition of a strong base to an aqueous solution of cations. The investigation of the samples that were quenched at different times after

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service