Skip to Content
Merck
All Photos(1)

Key Documents

909521

Sigma-Aldrich

Methyl 2,3,4,6-tetra-O-acetyl-α-D-glucopyranoside

≥95%

Synonym(s):

Tetraacetyl methyl glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O10
CAS Number:
Molecular Weight:
362.33
MDL number:

Assay

≥95%

form

powder

storage temp.

2-8°C

InChI

1S/C15H22O10/c1-7(16)21-6-11-12(22-8(2)17)13(23-9(3)18)14(24-10(4)19)15(20-5)25-11/h11-15H,6H2,1-5H3

InChI key

UYWUMFGDPBMNCA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service