907596
(4aR,6S,7R,8R,8aR)-2,2-di-tert-Butyl-6-(phenylthio)hexahydropyrano[3,2-d][1,3,2]dioxasiline-7,8-diol
≥95%
Synonym(s):
Phenyl 4,6-O-di-tert-butylsilyl-1-thio-β-D-galactoside
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
≥95%
form
powder
storage temp.
2-8°C
Application
This is a protected galactopyranoside useful as a building block for synthesis of complex carbohydrates. The compound has β-phenylthio and 4,6-ditert-butylsilyl protecting groups.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Carbohydrates and their biologically active glycoconjugate analogs are emerging as an important class of biomolecules.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service