Skip to Content
Merck
All Photos(1)

Key Documents

90667

Supelco

Flufenacet-(phenyl-d4)

PESTANAL®, analytical standard

Synonym(s):

Flufenacet-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14D4H9F4N3O2S
Molecular Weight:
367.36
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥97.5 atom % D

product line

PESTANAL®

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

CC(C)N(C(COC1=NN=C(C(F)(F)F)S1)=O)C2=C([2H])C([2H])=C(F)C([2H])=C2[2H]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service