Skip to Content
Merck
All Photos(1)

Documents

90452

Sigma-Aldrich

Triethylene glycol monohexyl ether

≥97.0% (GC)

Synonym(s):

C6E3, Hexyl triethylene glycol ether, Hexyltriglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5(OCH2CH2)3OH
CAS Number:
Molecular Weight:
234.33
Beilstein:
1757858
EC Number:
MDL number:
UNSPSC Code:
12161900

Assay

≥97.0% (GC)

refractive index

n20/D 1.442

density

0.961 g/mL at 20 °C

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCOCCOCCOCCO


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service