Skip to Content
Merck
All Photos(1)

Key Documents

89788

Millipore

V-Factor Disks

suitable for microbiology, for the presumptive identification of Haemophilus species

Synonym(s):

Coenzyme-diphosphopyridine-nucleotide Test Disks

Sign Into View Organizational & Contract Pricing

Select a Size

50 DISCS
€24.20

€24.20


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
50 DISCS
€24.20

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

€24.20


Please contact Customer Service for Availability

Request more information

Quality Level

sterility

sterile

product line

BioChemika

form

disc

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 50 discs

technique(s)

microbe id | growth & motility: suitable

application(s)

clinical testing
food and beverages

microbiology

storage temp.

2-8°C

suitability

Haemophilus spp. (species)

General description

V-Factor Discs are used for the presumptive identification of Haemophilus species based on their requirements for X or V growth factors or both. Members of the genus Haemophilus require accessory growth factors for in vitro growth. Some Haemophilus spp. require X factor (hemin) alone, V factor (nicotinamide adenine dinucleotide [NAD]) alone, or a combination of the two. Consequently, this significant difference in growth factor requirements of Haemophilus spp. allows for their differentiation. V factor comprises nicotinamide adenine dinucleotide (NAD) or nicotinamide adenine dinucleotide phosphate (NADP). The organisms will grow only around the disk that provides the appropriate factor for the growth of the organism. Differentiation is hence based on the presence or absence of growth around the discs impregnated with V-factor.

Application

V-Factor Discs are used for the presumptive identification of Haemophilus species and find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Packaging

one package contains 50 test discs (sterile filter paper impregnated with coenzyme-diphosphopyridine nucleotide)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service