Skip to Content
Merck
All Photos(1)

Key Documents

89122

Supelco

Gluconapin potassium salt monohydrate

analytical standard

Synonym(s):

1-S-[N-(Sulfooxy)-4-pentenimidoyl]-1-thio-β-D-glucopyranose monopotassium salt monohydrate, 3-Butenylglucosinolate potassium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18KNO9S2 · H2O
CAS Number:
Molecular Weight:
429.51
UNSPSC Code:
41116107

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service