Skip to Content
Merck
All Photos(1)

Key Documents

88789

Supelco

Formaldehyde dimethyl acetal

analytical standard

Synonym(s):

Dimethoxymethane, Methylal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(OCH3)2
CAS Number:
Molecular Weight:
76.09
Beilstein:
1697025
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.354

bp

41-43 °C

density

0.860 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COCOC

InChI

1S/C3H8O2/c1-4-3-5-2/h3H2,1-2H3

InChI key

NKDDWNXOKDWJAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formaldehyde dimethyl acetal is formed as a by-product during the production of trioxane, a crucial intermediate for the manufacture of polyacetal plastics.[1]

Application

2-Fluorobiphenyl may be used an analytical standard for the determination of the analyte in surface water samples, chemical raw materials, binary and ternary mixtures with methanol and/or water by various chromatography techniques.[1][2][3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Miermans et al.
Chemosphere, 40(1), 39-48 (2000-02-09)
Volatile organic compounds are widely introduced into the Dutch aquatic environment. Liquid-liquid extraction and isolation by means of resins give poor recoveries for volatiles. In this study, a method has been developed to analyse these compounds with a purge and
Vapor- liquid and liquid- liquid equilibria in binary and ternary mixtures of water, methanol, and methylal
Albert M, et al.
Journal of Chemical and Engineering Data, 46(4), 897-903 (2001)
Research on method for determination of methylal in chemical raw materials by head space GC-MS
ZHANG Y, et al.
Chinese journal of Health Laboratory Technology, 6 (2010)
Yuchuan Fu et al.
Chemical communications (Cambridge, England), (21)(21), 2172-2174 (2007-05-24)
Dimethoxymethane was synthesized from the direct oxidation of methanol with high conversion and selectivity over specially designed bifunctional V(2)O(5)/TiO2 catalysts with redox and enhanced acidic character, in which the surface acidity played an essential role for inhibiting the formation of
[Methylal: its metabolism and hygienic standardization in the air of work areas].
L A Tomilina et al.
Gigiena truda i professional'nye zabolevaniia, (6)(6), 27-29 (1984-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service