Skip to Content
Merck
All Photos(3)

Key Documents

86827

Millipore

Hybriscan I kit

Lactobacillus lindneri, suitable for environmental, food and beverages,microbiology and in situ hybriziation

Synonym(s):

Lactobacillus lindneri HybriScanI, FastScan Lactobacillus brevis

Sign Into View Organizational & Contract Pricing

Select a Size

48 TESTS
€290.00

€290.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
48 TESTS
€290.00

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

€290.00


Please contact Customer Service for Availability

Request more information

Product Name

HybriScanI Lactobacillus lindneri, suitable for food and beverages, microbiology, suitable for microbe id | in situ hybriziation

usage

sufficient for 48 reactions

manufacturer/tradename

(HybriScan)

technique(s)

microbe id | in situ hybriziation: suitable

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

Lactobacillus spp.

General description

HybriScan I kits are intended for the identification and result confirmation of microorganisms using rRNA as the detection target. Several customers also use it to detect microorganisms in food, beverage and water samples. The principle is based on in situ sandwich hybridization with two specific probes. As the rRNA of dead cells is degraded in the presence of RNase, only living cells are detected. Each cell has several thousand rRNA copies, so neither PCR nor a polymerase is needed for this technology, which means that this molecular biology test is not inhibited by the sample matrix.

Features and Benefits

  • Sensitivity (cfu/assay): 1000

Legal Information

HybriScan is a trademark of ScanBec GmbH

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Information about lactobacilli, rod-shaped, Gram-positive, fermentative, facultative anaerobic or microaerophilic organotrophs. The lactobacillus organtroph belongs to the lactic acid bacteria group.

HybriScan™ assays provide rapid, accurate molecular detection of beer spoilage microorganisms, ensuring excellent product quality in breweries.

Bifidobacteria-selective media detects beneficial bacteria, aiding quality control in microbiology labs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service