Skip to Content
Merck
All Photos(1)

Key Documents

86541

Sigma-Aldrich

Testosterone propionate

Testosterone propionate

tested according to Ph. Eur.

Synonym(s):

Testosteroni propionas, 17β-Hydroxy-4-androsten-3-one 17-propionate, 17β-Propionyloxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one 17-propionate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€86.20

€86.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€86.20

About This Item

Empirical Formula (Hill Notation):
C22H32O3
CAS Number:
Molecular Weight:
344.49
Beilstein:
3221760
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.21

€86.20


Please contact Customer Service for Availability

Request a Bulk Order

Agency

USP/NF
tested according to Ph. Eur.

form

solid

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])OC(=O)CC

InChI

1S/C22H32O3/c1-4-20(24)25-19-8-7-17-16-6-5-14-13-15(23)9-11-21(14,2)18(16)10-12-22(17,19)3/h13,16-19H,4-12H2,1-3H3/t16-,17-,18-,19-,21-,22-/m0/s1

InChI key

PDMMFKSKQVNJMI-BLQWBTBKSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Testosterone propionate is the esterified form of testosterone that is most commonly used in clinical applications for the treatment of hypogonadism, oligospermia, and impotence.[1] 17β-hydroxyl group of propionate is esterified to get this form. It is a fast and short-acting form when compared to enanthate and cypionate forms, which are long-acting esters more commonly used in androgen replacement therapies.[2]

Application

Testosterone propionate has been used:
  • to determine the effect of testosterone on the uptake of certain amino acids, such as isoleucine and α-(methylamino)isobutyric acid[3]
  • in a study to understand the effects of testosterone on brain aromatase activity in neonatal quail brain[4]
  • for testosterone treatment of Celf1 knockout mice in a study to investigate the upregulation of aromatase activity in association with hypogonadism[5]
  • to investigate the ontogeny of aromatase-expressing neurons in Japanese quail brain[6]
  • to study the effects of in vivo administration of testosterone propionate on fluid reabsorption in the efferent ducts of rat.[7]

Biochem/physiol Actions

Testosterone is the major circulating androgen in men and is mainly produced by the testis. A small amount is secreted by the adrenal gland. It is produced from cholesterol in the Leydig cells. Testosterone is metabolized to dihydrotestosterone, a potent androgen that associates with a cytoplasmic receptor protein, to form a complex. This complex is activated and transported into the nucleus, where it triggers several biological responses. It mainly participates in the development of the male phenotype during embryogenesis and at puberty.[8][9][10]

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Clinical Use and Abuse of Androgens and Antiandrogens
Becker KL., (eds.)
Principles and Practice of Endocrinology and Metabolism, 957-957 null
M Fujioka et al.
The Journal of clinical endocrinology and metabolism, 63(6), 1361-1364 (1986-12-01)
The pharmacokinetic characteristics of testosterone propionate were studied in normal men after a single im dose of 25 mg testosterone propionate-19,19,19-d3. Plasma levels of testosterone propionate-19,19,19-d3, its active metabolite testosterone-19,19,19-d3, and endogenous testosterone were measured by gas chromatography-mass spectrometry. Testosterone
L A Hansen et al.
International journal of andrology, 20(5), 265-273 (2005-09-01)
The regulation by oestradiol and testosterone of fluid reabsorption in the efferent ducts of the rat was investigated by determining the effects of administering the hormones and their antagonists. Untreated rats were compared to animals treated for 7 days with
J D Wilson et al.
Human genetics, 58(1), 78-84 (1981-01-01)
Male and female embryos develop in an identical fashion during the initial portion of gestation. If the indifferent gonad differentiates into an ovary (or if no gonad is present), a female phenotype is formed. Male phenotypic differentiation, however, requires the
Sylvia M Bardet et al.
Neuroreport, 21(5), 376-380 (2010-02-12)
It was shown earlier that, in Japanese quail the mechanism controlling the induction by testosterone of aromatase activity develops between embryonic days 10 and 14. The cellular processes underlying this activation have, however, not been investigated in detail. Here, we

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service