Skip to Content
Merck
All Photos(1)

Key Documents

86321

Supelco

Calibrator 0,0 for FOS (Food-Oil-Sensor)

86.5% isopropyl myristate basis, 13.5% heptadecane basis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12164500
NACRES:
NA.21

Assay

13.5% heptadecane basis
86.5% isopropyl myristate basis

form

liquid

application(s)

food and beverages
general analytical

Application

Standard for quality control of frying oils

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service