Skip to Content
Merck
All Photos(1)

Documents

85969

Supelco

Methoxypolyethylene glycol 5,000 propionic acid N-succinimidyl ester

≥80%

Synonym(s):

Polyethylene glycol, mono-Methyl polyethylene glycol 5′000 propionic acid N-succinimidyl ester, PEG-succinimidyl propionate, Polyethylene glycol N-succinimidyl propionate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥80%

form

solid

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service