Skip to Content
Merck
All Photos(1)

Key Documents

85000

Sigma-Aldrich

L-Serine methyl ester hydrochloride

purum, ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
155.58
Beilstein:
3559591
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

Assay

≥99.0% (AT)

optical activity

[α]20/D +5±0.5°, c = 2% in methanol

optical purity

enantiomeric ratio: ≥99.0:1.0 (GC)

mp

163 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

Cl.COC(=O)[C@@H](N)CO

InChI

1S/C4H9NO3.ClH/c1-8-4(7)3(5)2-6;/h3,6H,2,5H2,1H3;1H/t3-;/m0./s1

InChI key

NDBQJIBNNUJNHA-DFWYDOINSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Sélambarom et al.
Carbohydrate research, 330(1), 43-51 (2001-02-24)
The reaction of L-serine methyl ester hydrochloride (1) with paraformaldehyde (2) in dichloromethane in the presence of triethylamine afforded a novel compound: [lS,2S,6S,7S]-1,6-diaza-4,9-dioxa-2,7-dimethoxycarbonylbicyclo[4.4.1]undecane (4) as a 2:3 adduct of 1 with 2. 1H and 13C NMR spectroscopy were unable to
Cédric Couturier et al.
Organic letters, 8(10), 2183-2186 (2006-05-05)
[reaction: see text] Reaction of N,N-dibenzyl-O-methylsulfonyl serine methyl ester with a variety of heteronucleophiles (sodium azide, sodium phthalimide, amines, thiols) and carbanions (sodium malonate) gave, via an aziridinium intermediate, the corresponding beta-amino or alpha,beta-diamino ester in good to excellent yield.
Acylated serine derivatives: a unique class of arthropod pheromones of the Australian redback spider, Latrodectus hasselti.
Elena Jerhot et al.
Angewandte Chemie (International ed. in English), 49(11), 2037-2040 (2010-02-11)
Carlos Aydillo et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(17), 4840-4848 (2007-03-17)
A new chiral serine equivalent and its enantiomer have been synthesized from (S)- and (R)-N-Boc-serine methyl esters (Boc: tert-butyloxycarbonyl). The use of these compounds as chiral building blocks has been demonstrated in the synthesis of alpha-alkyl alpha-amino acids by diastereoselective
Yu Harayama et al.
Chemical communications (Cambridge, England), (13)(13), 1764-1766 (2005-03-26)
The use of hypervalent iodine(III) reagents allowed us to develop the novel and efficient direct synthesis of N,O-acetal compounds via the oxidative fragmentation reaction of alpha-amino acids or alpha-amino alcohols; furthermore, we succeeded in developing an improved synthesis of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service