Skip to Content
Merck
All Photos(1)

Key Documents

80901

Supelco

Lead (impurities)

manufactured by MBH Analytical Ltd (83X PR8)

Synonym(s):

Lead

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

material

disks

grade

analytical standard

resistivity

20.65 μΩ-cm

technique(s)

XRF: suitable

diam. × H

40 mm × 15 mm , dimensions are approximate

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

format

matrix material

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

Analysis Note

Values of analytes vary lot to lot.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenyi Yuan et al.
Journal of the Air & Waste Management Association (1995), 63(4), 418-423 (2013-05-22)
A novelprocess ofmechanochemical sulfidization for the treatment of cathode ray tube (CRT)funnel glass has been investigated by co-grinding with the element sulfur The ground samples were characterized by means of a set of analytical methods, including scanning electron microscopy (SEM)
James D Blando et al.
Journal of environmental health, 75(9), 20-27 (2013-06-06)
The U.S. Environmental Protection Agency recently implemented the Renovation, Repair and Painting (RRP) rule that applies to pre-1978 residences because of the potential presence of lead-based paint. Enforcement of this rule may be difficult and therefore it is crucial to
Nattaporn Youravong et al.
International dental journal, 63(3), 123-129 (2013-05-23)
Saliva has been suggested as a useful biomarker of environmental contamination, including exposure to lead. The study examined the associations between salivary lead level and the following factors: blood lead level, the number of decayed and filled surfaces (dfs) on
Kai-Ying Chiang et al.
Journal of environmental sciences (China), 25(3), 626-636 (2013-08-08)
This study investigates biogeochemical reductive release of arsenate from beudantite into solution in a crater area in northern Taiwan, using a combination of X-ray absorption near-edge structure (XANES) and atomic absorption spectrometry. Total arsenic (As) concentrations in the soil were
Karsten Witt et al.
Brain : a journal of neurology, 136(Pt 7), 2109-2119 (2013-06-27)
Deep brain stimulation of the subthalamic nucleus improves motor functions in patients suffering from advanced Parkinson's disease but in some patients, it is also associated with a mild decline in cognitive functioning about one standard deviation from the preoperative state.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service