Skip to Content
Merck
All Photos(1)

Key Documents

79840

Sigma-Aldrich

N-Phthaloyl-L-glutamic acid

purum, ≥98.0% (T)

Synonym(s):

PhGA

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$33.35
250 MG
$46.00
1 G
$111.55
5 G
$377.20

Vendor SKU: A253042-100MG

$33.35


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$33.35
250 MG
$46.00
1 G
$111.55
5 G
$377.20

About This Item

Empirical Formula (Hill Notation):
C13H11NO6
CAS Number:
Molecular Weight:
277.23
Beilstein:
90136
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Vendor SKU: A253042-100MG

$33.35


Ships in 7 days - from Aldrich Partner.

grade

purum

Assay

≥98.0% (T)

optical activity

[α]20/D −48±2°, c = 3% in dioxane

mp

160-162 °C (lit.)
160-162 °C

application(s)

peptide synthesis

SMILES string

OC(=O)CC[C@H](N1C(=O)c2ccccc2C1=O)C(O)=O

InChI

1S/C13H11NO6/c15-10(16)6-5-9(13(19)20)14-11(17)7-3-1-2-4-8(7)12(14)18/h1-4,9H,5-6H2,(H,15,16)(H,19,20)/t9-/m0/s1

InChI key

FEFFSKLJNYRHQN-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

N-protected derivative of L-glutamic acid. The anhydride (prepared with DCCI or Ac2O without racemization) is used for γ-L-glutamylations[1][2][3][4]; The phthaloyl protection can be removed mildly[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and structure of coprine, the in vivo aldehyde dehydrogenase inhibitor in Coprinus atramentarius; Syntheses of coprine and related cyclopropa4one derivatives.
P Lindberg et al.
Journal of the Chemical Society. Perkin transactions 1, (6), 684-691 (1977-01-01)
J.O. Osby et al.
Tetrahedron Letters, 25, 2093-2093 (1984)
Agaridoxin, a mushroom metabolite. Isolation, structure, and synthesis.
A Szent-Gyorgyi et al.
The Journal of organic chemistry, 41(9), 1603-1606 (1976-04-30)
-Glutamyl cyclotransferase. Distribution, isozymic forms, and specificity.
M Orlowski et al.
The Journal of biological chemistry, 248(8), 2836-2844 (1973-04-25)
U. Nagai et al.
Tetrahedron Letters, 26, 647-647 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service