Skip to Content
Merck
All Photos(2)

Key Documents

79589

Supelco

Phloridzin dihydrate

analytical standard

Synonym(s):

1-[2-(β-D-Glucopyranosyloxy)-4,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)-1-propanone, Phloretin 2′-β-D-glucopyranoside, Phloretin 2′-β-D-glucoside, Phlorizin dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O10 · 2H2O
CAS Number:
Molecular Weight:
472.44
Beilstein:
66621
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

7.6% water (dihydrate, theroy)

mp

113-114 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

O.O.OC[C@H]1O[C@@H](Oc2cc(O)cc(O)c2C(=O)CCc3ccc(O)cc3)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H24O10.2H2O/c22-9-16-18(27)19(28)20(29)21(31-16)30-15-8-12(24)7-14(26)17(15)13(25)6-3-10-1-4-11(23)5-2-10;;/h1-2,4-5,7-8,16,18-24,26-29H,3,6,9H2;2*1H2/t16-,18-,19+,20-,21-;;/m1../s1

InChI key

XQWBNXSENPTIDY-YXMARJSJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

Used to induce experimental glycosuria.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongxi Li et al.
Molecules (Basel, Switzerland), 24(1) (2018-12-28)
The present study examined the effect of Lactobacillus plantarum ATCC14917 fermentation on the chemical composition and antioxidant activity of apple juice. Apple juice was fermented and examined of its antioxidant activity using chemical models and cellular antioxidant assay. Furthermore, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service