Skip to Content
Merck
All Photos(2)

Key Documents

78645

Supelco

Levetiracetam

analytical standard

Synonym(s):

(αS)-α-Ethyl-2-oxo-1-pyrrolidineacetamide, 2(S)-(2-Oxopyrrolidin-1-yl)butyramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O2
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

optical activity

[α]/D -90±5°, c = 1 in acetone

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

CC[C@H](N1CCCC1=O)C(N)=O

InChI

1S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)/t6-/m0/s1

InChI key

HPHUVLMMVZITSG-LURJTMIESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Levetiracetam is an orally active antiepileptic drug, having a high therapeutic index and potential antiepileptogenic and anticonvulsant effects.

Application

Levetiracetam may be used as a reference standard in the determination of levetiracetam in biological fluid samples using high-performance liquid chromatography coupled with electrospray tandem mass spectrometry (HPLC-ESI-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Levetiracetam is a pyrrolidine with antiepileptic activity.
Levetiracetam is a pyrrolidine with antiepileptic activity. Stereoselective binding of levetiracetam was confined to synaptic plasma membranes in the central nervous system with no binding occurring in peripheral tissue. Levetiracetam inhibits burst firing without affecting normal neuronal excitability, which suggests that it may selectively prevent hyper-synchronization of epileptiform burst firing and propagation of seizure activity.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of levetiracetam, a novel antiepileptic drug, on convulsant activity in two genetic rat models of epilepsy.
Gower T.A, et al.
Epilepsy Research, 22(3), 207-213 (1995)
Determination of levetiracetam in human plasma/serum/saliva by liquid chromatography-electrospray tandem mass spectrometry.
Guo T, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 375(1-2), 115-118 (2007)
Pharmacokinetic profile of levetiracetam: toward ideal characteristics.
Patsalos, PN
Pharmacology & Therapeutics, 85(2), 77-85 (2000)
Jingdong Chao et al.
Advances in therapy, 32(3), 270-283 (2015-03-17)
The purpose of the present study was to investigate the traceability of adverse events (AEs) for branded and generic drugs with identical nonproprietary names and to consider potential implications for the traceability of AEs for branded and biosimilar biologics. Adverse
Krzysztof Sendrowski et al.
Pharmacological reports : PR, 67(2), 326-331 (2015-02-26)
Alzheimer's disease (AD) is a common neurodegenerative disorder, in which progressive neuron loss, mainly in the hippocampus, is observed. The critical events in the pathogenesis of AD are associated with accumulation of β-amyloid (Aβ) peptides in the brain. Deposits of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service