Skip to Content
Merck
All Photos(2)

Key Documents

77960

Supelco

Ginsenoside Re

analytical standard

Synonym(s):

(3β,6α,12β)-20-(β-D-Glucopyranosyloxy)-3,12-dihydroxydammar-24-en-6-yl 2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranoside, Chikusetsusaponin IVc, Ginsenoside B2, NSC 308877, Panaxoside Re, Sanchinoside Re

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€278.00

€278.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€278.00

About This Item

Empirical Formula (Hill Notation):
C48H82O18
CAS Number:
Molecular Weight:
947.15
UNSPSC Code:
85151701
NACRES:
NA.24

€278.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2O[C@H]3C[C@]4(C)[C@H](C[C@@H](O)[C@@H]5[C@H](CC[C@@]45C)[C@](C)(CC\C=C(/C)C)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)[C@@]7(C)CC[C@H](O)C(C)(C)[C@H]37)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C48H82O18/c1-21(2)11-10-14-48(9,66-42-38(60)35(57)32(54)26(19-49)63-42)23-12-16-46(7)30(23)24(51)17-28-45(6)15-13-29(52)44(4,5)40(45)25(18-47(28,46)8)62-43-39(36(58)33(55)27(20-50)64-43)65-41-37(59)34(56)31(53)22(3)61-41/h11,22-43,49-60H,10,12-20H2,1-9H3/t22-,23-,24+,25-,26+,27+,28?,29-,30+,31-,32+,33+,34+,35?,36-,37+,38+,39+,40-,41-,42-,43+,45+,46+,47+,48-/m0/s1

InChI key

PWAOOJDMFUQOKB-QVPADXGFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Peng et al.
Cardiovascular therapeutics, 30(4), e183-e188 (2011-09-03)
Ginsenosides are the bioactive constituents of ginseng, a key herb in traditional Chinese medicine. As a single component of ginseng, ginsenoside Re (G-Re) belongs to the panaxatriol group. Many reports demonstrated that G-Re possesses the multifaceted beneficial pharmacological effects on
Yongjian Xiong et al.
Journal of natural medicines, 68(3), 530-538 (2014-03-25)
Ginsenoside Re (GRe) exerts diverse effects. Based on our observations, the present study was designed to investigate GRe-exerted bidirectional regulation (BR) on the contractility of isolated jejunal segment. Six pairs of different low and high contractile states of rat jejunal
Suporn Sukrittanon et al.
Life sciences, 115(1-2), 15-21 (2014-09-23)
Ginsenosides, active components in ginseng, have been shown to increase nitric oxide (NO) production in aortic endothelial cells. This effect was reversed by tetraethylammonium (TEA) inhibition of endothelial Ca(2+)-activated K(+) (KCa) channels. The objectives of this study, therefore, were to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service