Skip to Content
Merck
All Photos(1)

Key Documents

77663

Supelco

Terpineol

mixture of isomers, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€52.10
5 ML
€253.00

€52.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€52.10
5 ML
€253.00

About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€52.10


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

α-terpineol, 60.0-85.0% GC
β-terpineol, 1.0-15.0% GC
γ-terpineol, 10.0-25.0% GC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.483-1.485

bp

213-218 °C (lit.)

density

0.934 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=C)C1CCC(C)(O)CC1.CC2=CCC(CC2)C(C)(C)O

InChI

1S/2C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11;1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;9,11H,1,4-7H2,2-3H3

InChI key

MGABGZGUMNDCSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han Fu et al.
Nanoscale research letters, 9(1), 631-631 (2014-12-19)
Fabricating functional compounds on substrates with complicated morphology has been an important topic in material science and technology, which remains a challenging issue to simultaneously achieve a high growth rate for a complex nanostructure with simple controlling factors. Here, we
Jin-Wook Lee et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 15(12), 2595-2603 (2014-05-28)
A dichlorobenzene-functionalized hole-transporting material (HTM) is developed for a CH3NH3PbI3-based perovskite solar cell. Notwithstanding the similarity of the frontier molecular orbital energy levels, optical properties, and hole mobility between the functionalized HTM [a polymer composed of 2'-butyloctyl-4,6-dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate (TT-BO), 3',4'-dichlorobenzyl-4,6-dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate (TT-DCB)
Hyun-Woo Kang et al.
Faraday discussions, 176, 287-299 (2014-11-20)
The effect of double blocking layers on the Sb(2)S(3)-sensitized all solid state solar cell are investigated. Thin layers of ZrO(2) (blocking layer 1, BL1) and ZnS (blocking layer 2, BL2) are introduced at the TiO(2)/Sb(2)S(3) and Sb(2)S(3)/hole transporting material (HTM)
Barbara Sgorbini et al.
Journal of chromatography. A, 1376, 9-17 (2014-12-30)
Herbs and spices are used worldwide as food flavoring, thus determination of their identity, origin, and quality is mandatory for safe human consumption. An analysis strategy based on separative (HS-SPME-GC-MS) and non-separative (HS-SPME-MS) approaches is proposed for the volatile fraction
Karine Rech Begnini et al.
Journal of medicinal food, 17(10), 1129-1133 (2014-09-18)
Cancer is a leading cause of death and is responsible for one in eight deaths worldwide. The use of herbs as complementary medicine for cancer, especially advanced cancer, has recently increased. The aim of this study was to evaluate in

Protocols

GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service