Skip to Content
Merck
All Photos(1)

Key Documents

76552

Supelco

Polystyrene (low molecular) Standard ReadyCal Set M(p) ~250-65000

for GPC

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€605.00

€605.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
€605.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€605.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

storage temp.

2-8°C

Components

ReadyCal Standards are polymer cocktails pre-weighed into autosampler vials for the convenient calibration of GPC/SEC/GFC systems.
This kit contains 10 sets of 3 color-coded vials, each containing four polystyrenes of different molar masses. Molar masses range from ~266 to ~66,000 Da and are carefully selected so that polymer traces are baseline separated.
Sufficient material provided for at least 10 calibrations.
Suitable for calibrations in THF, toluene, DMF, DMAc, chloroform, TCB, DCB and NMP.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Barbara De Meester et al.
Nature communications, 11(1), 5020-5020 (2020-10-08)
Lignin causes lignocellulosic biomass recalcitrance to enzymatic hydrolysis. Engineered low-lignin plants have reduced recalcitrance but often exhibit yield penalties, offsetting their gains in fermentable sugar yield. Here, CRISPR/Cas9-generated CCR2(-/*) line 12 poplars have one knockout CCR2 allele while the other

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service