Skip to Content
Merck
All Photos(1)

Key Documents

74886

Supelco

Sodium 1-octanesulfonate solution

suitable for ion pair chromatography, LiChropur, concentrate, ampule

Synonym(s):

1-Octanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH2SO3Na
CAS Number:
Molecular Weight:
216.27
Beilstein:
3574241
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21
To inquire about this product 74886, please contact your local Merck office or dealer. Contact Technical Service

Quality Level

form

liquid

quality

LiChropur
ampule

packaging

ampule of 25 mL

technique(s)

ion pair chromatography: suitable

λ

in concentrate

UV absorption

λ: 210 nm Amax: 0.08
λ: 220 nm Amax: 0.05
λ: 230 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

[Na+].CCCCCCCCS([O-])(=O)=O

InChI

1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

HRQDCDQDOPSGBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Concentrate for 6x1 L ready-to-use solution. Dilute contents of one ampule (~0.3 M) to 1 L with HPLC grade water to obtain a 0.005 M eluent solution.
Sodium 1-octanesulfonate monohydrate (NaOS) is an ion-pairing reagent.[1]

Application

NaOS has been used as ion-pairing reagent for ultra-high performance liquid chromatography (UPLC) analysis with photodiode array detection to estimate paraquat (methyl viologen) in tissue homogenate.[1] It may also be used in determination of dopamine and its metabolites in brain microdialysates using liquid chromatography combined with electrospray ionization tandem mass spectrometry (LC–ESI-MS/MS).[2]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and simple analysis of paraquat in tissue homogenate by ultra-high performance liquid chromatography.
Merritt, T. J. S., et al.
Analytical Methods : Advancing Methods and Applications, 3.6, 1428-1432 (2011)
Kamila Syslová et al.
Journal of chromatography. A, 1218(21), 3382-3391 (2011-03-15)
A sensitive assay method was developed for a parallel, rapid and precise determination of dopamine and its metabolites, homovanillic acid, 3-methoxytyramine and 3,4-dihydroxyphenylacetic acid, from brain microdialysates. The method consisted of a pre-treatment step, freeze-drying (lyophilization), to concentrate dopamine and
J G Foulks et al.
Canadian journal of physiology and pharmacology, 63(10), 1327-1334 (1985-10-01)
The potency of various types of alkyl amphipathic (cationic, anionic, and neutral) as well as tertiary amine local anesthetics in impairing the excitability of frog skeletal muscle was markedly enhanced by an increase in temperature from 20 to 30 degrees
H T Jung et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(4), 1353-1357 (2001-02-15)
Equilibrium unilamellar vesicles are stabilized by one of two distinct mechanisms depending on the value of the bending constant. Helfrich undulations ensure that the interbilayer potential is always repulsive when the bending constant, K, is of order k(B)T. When K
Can Zhang et al.
Biomaterials, 29(9), 1233-1241 (2007-12-21)
Paclitaxel (Taxol), PTX) is a promising anti-cancer drug and has been successfully used to treat a wide variety of cancers. Unfortunately, serious clinical side effects are associated with it, which are caused by PTX itself and non-aqueous vehicle containing Cremophor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service