Skip to Content
Merck
All Photos(1)

Documents

74294

Sigma-Aldrich

Chloroethane

≥99.8%

Synonym(s):

Ethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5Cl
CAS Number:
Molecular Weight:
64.51
Beilstein:
1730751
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

vapor density

2.22 (vs air)

vapor pressure

19.2 psi ( 20 °C)

Assay

≥99.8%

impurities

≤0.02% water

bp

12.3 °C (lit.)

mp

−139 °C (lit.)

density

0.921 g/mL at 0-4 °C (lit.)

SMILES string

CCCl

InChI

1S/C2H5Cl/c1-2-3/h2H2,1H3

InChI key

HRYZWHHZPQKTII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroethane is a haloalkane used as an intermediate or as a solvent in organic synthesis. It has major applications in the production of tetraethyl lead, ethyl-cellulose, and an antiknock additive for gasoline. It is also used in cleaning and degreasing applications.

Application

Chloroethane can be used to prepare:
  • pyrido[1,2-a]pyrimidinone derivatives
  • 2H-perovskite compound [Me3NCH2CH3]CdCl3
  • benzo[cd]indol-2(1H)-ones derivatives as potent BRD4 inhibitors

Packaging

cylinder with 11.4 kg (net)

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective formation of chloroethane by the hydrochlorination of ethene using zinc catalysts
Conte M, et al.
J. Catal., 252(1), 23-29 (2007)
First discovery of novel pyrido [1,2-a] pyrimidinone mesoionic compounds as antibacterial agents
Liu D, et al.
Journal of Agricultural and Food Chemistry, 67(43), 11860-11866 (2019)
Design, synthesis and biological evaluation of benzo [cd] indol-2 (1H)-ones derivatives as BRD4 inhibitors
Feng Y, et al.
European Journal of Medicinal Chemistry, 152(16), 264-273 (2018)
Reversible high temperature dielectric switching in a 2 H-perovskite compound:[Me 3 NCH 2 CH 3] CdCl 3
Sun D, et al.
CrystEngComm, 21(16), 2669-2674 (2019)
A Banerjee et al.
Journal of dentistry, 38(6), 475-479 (2010-03-13)
Compare clinical effectiveness of sodium bicarbonate and bioactive glass powders used for dental prophylaxis. 25 patients were allocated to either good or poor oral hygiene subgroups (n=50). Using a double-blind, split-mouth model, all patients underwent prophylaxis treatment on mandibular teeth;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service