Skip to Content
Merck
All Photos(1)

Key Documents

74088

Supelco

Methomyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€181.00

€181.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€181.00

About This Item

Empirical Formula (Hill Notation):
C5H10N2O2S
CAS Number:
Molecular Weight:
162.21
Beilstein:
3081564
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€181.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CNC(=O)O\N=C(/C)SC

InChI

1S/C5H10N2O2S/c1-4(10-3)7-9-5(8)6-2/h1-3H3,(H,6,8)/b7-4+

InChI key

UHXUZOCRWCRNSJ-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Methomyl is a broad-spectrum insecticide that belongs to the family of oxime carbamate pesticides. It controls insects and nematode pests by inhibiting the activity of the enzyme acetylcholinesterase and preventing the hydrolysis of neurotransmitter acetylcholine.(1)
Methomyl has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin

As per Regulation (EC) No 1107/2009, repealing the directive 91/414, methomyl is not approved for use in the European Union (EU). A maximum residue limit (MRL) of 0.01 mg/kg is applicable for its presence in most of the commodities with an MRL of 0.2 mg/kg particularly for lettuce and cotton seeds, in accordance with Regulation (EU) 2016/1822.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The methomyl CRM may also find the following uses:


  • Detection of methomyl in food matrices (wheat and rice flours) using terahertz time-domain spectroscopy (THz-TDS)
  • Degradation of methomyl with intensifying agents such as hydrogen peroxide, Fenton reagent, and ozone by using hydrodynamic cavitation reactor (HC)
  • Degradation of methomyl by using the ultrasound cavitation (US) in combination with hydrogen peroxide, Fenton, and photo-Fenton process
  • Determination of methomyl using a bare boron-doped diamond electrode by voltammetric methods
  • Degradation of methomyl by the combination of Aminobacter sp. MDW-2 and Afipia sp. MDW-3

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service