Skip to Content
Merck
All Photos(1)

Key Documents

73082BC

Millipore

GDS Rotor-Gene Q Laptop International Power Cord

BioControl, For use with GDS, European adaptor

Synonym(s):

Laptop Power Cord Adaptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.85
Product 73082BC is not currently sold in your country. Contact Technical Service

manufacturer/tradename

BioControl Systems 73082EU

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

General description

The GDS Rotor-Gene® Q Laptop is for use with the Assurance® GDS Rotor-Gene® Q Thermocycler. The International Power Cord is configured for the EU region.

Application

GDS Rotor-Gene® Q Laptop International Power Cord is used to connect the Assurance® GDS Q laptop to a power source in the EU region.

Components

One international power adapter

Testing Method

PCR

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Rotor-Gene is a registered trademark of Qiagen GmbH

required but not provided

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service