Skip to Content
Merck
All Photos(1)

Key Documents

71680

Sigma-Aldrich

Sodium bitartrate monohydrate

purum p.a., ≥98% (T)

Synonym(s):

L-(+)-Tartaric acid monosodium salt, Sodium hydrogen tartrate monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€61.90
1 KG
€212.00

€61.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€61.90
1 KG
€212.00

About This Item

Empirical Formula (Hill Notation):
C4H5NaO6 · H2O
CAS Number:
Molecular Weight:
190.08
Beilstein:
6099125
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
form:
crystals

€61.90


Please contact Customer Service for Availability

Request a Bulk Order

grade

purum p.a.

Quality Level

Assay

≥98% (T)

form

crystals

optical activity

[α]20/D +21±2°, c = 1% in H2O

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

functional group

carboxylic acid

SMILES string

[Na+].[H]O[H].OC(C(O)C([O-])=O)C(O)=O

InChI

1S/C4H6O6.Na.H2O/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);;1H2/q;+1;/p-1

InChI key

LLVQEXSQFBTIRD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bitartrate monohydrate (SBTMH) is an organometallic compound having nonlinear optical properties. Its synthesis at ambient temperature has been reported.[1]

Application

  • Radiation sensitivity of tartrate compounds: A study investigated the radiation sensitivity of various tartrate compounds, including Sodium bitartrate monohydrate, to understand their potential in radiation processing applications. This research contributes to optimizing the use of Sodium bitartrate monohydrate in radiation-related chemical processes (Bal and Tuner, 2014).

  • Reclaiming Agriceuticals from Sweet potato (Ipomoea batatas [L.] Lam.) By-Products: This study highlights the potential of utilizing agricultural by-products rich in lutein, enhancing sustainability and resource efficiency in pharma biotech industries. The research underlines the significant bioavailability of lutein extracted from natural sources, advancing its application in nutraceuticals and functional foods (Liu T et al., 2024).

  • The Potential of Natural Carotenoids-Containing Sericin of the Domestic Silkworm Bombyx mori: Investigates the use of sericin, enriched with natural carotenoids including lutein, for its potential in skin care and pharmaceutical applications, demonstrating the synergy between biotechnology and traditional industries (Liu S et al., 2024).

  • Chemical Profile Characterization of Fruit and Vegetable Juices after Fermentation with Probiotic Strains: This article explores the enhancement of antioxidant properties of juices through fermentation, noting an increase in bioavailable lutein, which is crucial for expanding the use of lutein in dietary supplements and functional beverages (Mantzourani I et al., 2024).

  • Valorisation of sea urchin (Paracentrotus lividus) gonads through canning: Describes a process to enhance the nutritional value of sea urchin gonads, including the stabilization of lutein, facilitating its use in the pharmaceutical and food industries for improved eye and skin health (Camacho C et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and growth of sodium bitartrate monohydrate a new organometallic nonlinear optical single crystal.
Shankar VS, et al.
Current Applied Physics, 9(5), 1125-1128 (2009)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service