Skip to Content
Merck
All Photos(1)

Key Documents

71287

Sigma-Aldrich

Sodium (meta)arsenite

purum p.a., ≥99.0% (RT)

Synonym(s):

Sodium dioxoarsenate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaAsO2
CAS Number:
Molecular Weight:
129.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

purum p.a.

Assay

≥99.0% (RT)

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].[O-][As]=O

InChI

1S/AsHO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

PTLRDCMBXHILCL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lian-Qun Qiu et al.
Environmental health perspectives, 123(4), 324-330 (2014-12-11)
Inorganic arsenic species are potent environmental toxins and causes of numerous health problems. Most studies have assumed that arsenic-induced changes in mRNA levels result from effects on gene transcription. We evaluated the prevalence of changes in mRNA stability in response
Ranya A Amer et al.
BioMed research international, 2015, 981829-981829 (2015-08-15)
Coastal environments worldwide are threatened by the effects of pollution, a risk particularly high in semienclosed basins like the Mediterranean Sea that is poorly studied from bioremediation potential perspective especially in the Southern coast. Here, we investigated the physical, chemical
Haitian Lu et al.
Toxicological sciences : an official journal of the Society of Toxicology, 112(2), 363-373 (2009-09-22)
Rodent models have been extensively utilized to identify putative human immunotoxicants; however, even when immunotoxicity is established, uncertainty remains whether the effects are predictive of human risk. Therefore, the objective of this study was to establish a polyclonal immunoglobulin M
Beibei Cai et al.
Apoptosis : an international journal on programmed cell death, 13(6), 803-810 (2008-05-10)
Sodium arsenite induces apoptosis in PC12 cells by activating the stress-activated p38 MAP kinase and the pro-apoptotic Bcl-2 family protein Bim(EL). However, the relationship between p38 and Bim(EL) in this apoptosis has not been fully defined. Here, we report that
Hong Anh Duong et al.
Environmental science. Processes & impacts, 17(11), 1941-1951 (2015-10-10)
A simple and inexpensive method for the determination of various ionic species in different water matrices is discussed in this study. The approach is based on the employment of in-house-made capillary electrophoresis (CE) instruments with capacitively coupled contactless conductivity detection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service