Skip to Content
Merck
All Photos(1)

Key Documents

69168

Supelco

α-Methylstyrene monomer

analytical standard

Synonym(s):

2-Phenylpropene, Isopropenylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(CH3)=CH2
CAS Number:
Molecular Weight:
118.18
Beilstein:
969405
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

contains

~0.0025% 4-tert-butylcatechol as stabilizer

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.539

bp

164-168 °C (lit.)

density

0.909 g/mL at 20 °C

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

neat

storage temp.

2-8°C

SMILES string

CC(=C)c1ccccc1

InChI

1S/C9H10/c1-8(2)9-6-4-3-5-7-9/h3-7H,1H2,2H3

InChI key

XYLMUPLGERFSHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

α-Methylstyrene may be used as an analytical reference standard for the determination of the analyte in workplace ambient air by gas chromatography-mass spectrometric optimization method.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Gas chromatography- mass spectroscopy optimization by computer simulation, application to the analysis of 93 volatile organic compounds in workplace ambient air.
Randon J, et al.
Analytica Chimica Acta, 812, 258-264 (2014)
D B Dzhusupova et al.
Mikrobiologiia, 54(1), 136-140 (1985-01-01)
The work was aimed at studying alpha-methylstyrene oxidation by three active Pseudomonas cultures. P. acidovorans 9 oxidized alpha-methylstyrene only to acetophenone whereas two P. aeruginosa cultures assimilated the compound entirely. Analysis of the intermediate products and enzymes catalyzing the aromatic
A N Ilialetdinov et al.
Mikrobiologiia, 52(4), 609-614 (1983-07-01)
Two bacterial strains assimilating alpha-methylsterene as a sole carbon and energy source were isolated from the sewage of an industrial plant producing synthetic rubber. The morphological, cultural and physiologo-biochemical properties of the strains were studied and their taxonomic position was
[Kallikrein-kinin system indices in neurocirculatory dystonia patients who work in synthetic rubber manufacture].
N I Al'berton et al.
Gigiena truda i professional'nye zabolevaniia, (12)(12), 23-26 (1981-12-01)
National Toxicology Program technical report series, (543)(543), 1-210 (2008-08-08)
alpha-Methylstyrene is used in the production of acrylonitrile-butadiene-styrene resins and copolymers, which improve the impact and heat-resistant properties of polymers, specialty grades of plastics, rubber, and protective coatings. alpha-Methylstyrene also moderates polymerization rates and improves product clarity in coatings and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service