Skip to Content
Merck
All Photos(1)

Key Documents

68554

Supelco

γ-Hexalactone

analytical standard

Synonym(s):

γ-Caprolactone, γ-Ethyl-γ-butyrolactone

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€90.00

€90.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€90.00

About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
Beilstein:
6114822
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€90.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.437-1.441
n20/D 1.439 (lit.)

bp

219 °C (lit.)

density

1.023 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCC1CCC(=O)O1

InChI

1S/C6H10O2/c1-2-5-3-4-6(7)8-5/h5H,2-4H2,1H3

InChI key

JBFHTYHTHYHCDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-Hexalactone is a volatile flavoring compound largely present in food and beverage products, fruits, vegetables, etc.[1][2][3][4][5]

Application

γ-Hexalactone may be used as an analytical standard for the determination of the analyte in wine,[1] fruits,[2] biological samples,[6] meat products,[3] and e-cigarette refill solutions[4] by various chromatography techniques.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of Potent Aroma Compounds of Cape Gooseberry (Physalis Peruviana L.) Fruits Grown in Antalya Through the Determination of Odor Activity Values
Yilmaztekin, Murat
International journal of food properties, 17, 469-480 (2013)
Xiaoxia Shi et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 913-914, 69-76 (2012-12-29)
The "enzymatic hydrolysis-mild thermal oxidation" method was employed to obtain oxidized tallow. Nine beeflike flavours (BFs) were prepared through Maillard reaction with oxidized tallow and other ingredients. Volatile compounds of oxidized tallow and beeflike flavours were analysed by SPME/GC-MS. Six
Shuang Chen et al.
Journal of agricultural and food chemistry, 61(47), 11295-11302 (2013-10-09)
The aroma profile of Chinese rice wine was investigated in this study. The volatile compounds in a traditional Chinese rice wine were extracted using Lichrolut EN and further separated by silica gel normal phase chromatography. Seventy-three aroma-active compounds were identified
Justyna Aszyk et al.
Journal of chromatography. A, 1519, 45-54 (2017-09-04)
Liquid chromatography-tandem mass spectrometry with electrospray ionization (HPLC-ESI-MS/MS) methods were developed for the simultaneous determination of 42 flavouring compounds and nicotine in liquids for e-cigarettes. The chromatographic separation was performed using an Ace® Ultracore™ SuperC18™ (100×2.1mm, 2.5μm) column in both
Carlo Bicchi et al.
Journal of chromatography. A, 1212(1-2), 114-123 (2008-10-31)
The analysis of complex real-world samples of vegetable origin requires rapid and accurate routine methods, enabling laboratories to increase sample throughput and productivity while reducing analysis costs. This study examines shortening enantioselective-GC (ES-GC) analysis time following the approaches used in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service