Skip to Content
Merck
All Photos(2)

Key Documents

68277

Supelco

Palmitic-d31 acid

analytical standard

Synonym(s):

Palmitic acid-d31, Hexadecanoic-d31 acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€75.20

€75.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€75.20

About This Item

Linear Formula:
CD3(CD2)14CO2H
CAS Number:
Molecular Weight:
287.62
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€75.20


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

isotopic purity

≥98 atom % D

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

58-60 °C
61-64 °C

application(s)

food and beverages

format

neat

functional group

carboxylic acid

mass shift

M+31

shipped in

ambient

storage temp.

room temp

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2

InChI key

IPCSVZSSVZVIGE-SAQPIRCFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service