Skip to Content
Merck
All Photos(1)

Key Documents

68122

Millipore

Salmonella ID Membrane

suitable for microbiology, white colored, sterile, diameter 70 mm

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

10 DISCS
€74.30

€74.30


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
10 DISCS
€74.30

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

€74.30


Please contact Customer Service for Availability

Request more information

membrane size

70 mm

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Salmonella spp.

General description

Salmonella ID Membrane is a substrate-carrying ID membrane used for rapid identification and differentiation of Salmonella sp. from coliforms through characteristic color reactions. The membranes contain chromogenic substrates, which serve as the basis for microbe differentiation by color. The principle is based on a specific substrate enzyme reaction in which Salmonella organisms will fluoresce light purple to blue color when visualized under UV light.

Application

Salmonella ID Membrane is used for rapid detection, confirmation, and differentiation of Salmonella sp. from other coliforms in water and food samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Escherichia coli (25922) - blueSalmonella Typhimurium (14028) - light purpleSalmonella Enteritidis (13076) - light purpleKlebsiella pneumoniae (13883) - colorless, mucoid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service