Skip to Content
Merck
All Photos(1)

Key Documents

65369

Supelco

(S)-(−)-α-Methoxy-α-trifluoromethylphenylacetic acid

for chiral derivatization, LiChropur, ≥99.0%

Synonym(s):

(−)-MTPA, Mosher’s acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9F3O3
CAS Number:
Molecular Weight:
234.17
Beilstein:
4684048
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

Quality Level

Assay

≥99.0% (T)
≥99.0%

form

solid

optical activity

[α]20/D −73±1°, c = 2% in methanol

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

quality

LiChropur

technique(s)

HPLC: suitable

bp

90 °C/0.1 mmHg

mp

46-49 °C (lit.)

storage temp.

2-8°C

SMILES string

CO[C@](C(O)=O)(c1ccccc1)C(F)(F)F

InChI

1S/C10H9F3O3/c1-16-9(8(14)15,10(11,12)13)7-5-3-2-4-6-7/h2-6H,1H3,(H,14,15)/t9-/m0/s1

InChI key

JJYKJUXBWFATTE-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Determination of absolute configuration of stereogenic carbinol carbons; doi:10.1038/nprot.2007.354[1]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Sui et al.
The Journal of organic chemistry, 75(9), 2942-2954 (2010-04-17)
Two different mixture synthesis routes have been used to make the four stereoisomers of petrocortyne A. A first quick and dirty route provided a mixture of the four isomers in nonselective fashion. Mosher and 2-naphthylmethoxyacetic acid (NMA) ester methods were
D B Matthews et al.
Journal of chromatography. B, Biomedical sciences and applications, 695(2), 279-285 (1997-08-01)
Methods for the nuclear magnetic resonance and gas chromatographic analysis of the enantiomers of p-trifluoromethylmandelic acid (p-TFM) and Mosher's acid (alpha-methoxy-alpha-(trifluoromethyl)phenylacetic acid) present in rat urine samples are described. Gas chromartography was performed using cyclodextrin capillary columns with both compounds
J Martín Torres-Valencia et al.
Phytochemical analysis : PCA, 13(6), 329-332 (2002-12-24)
A method to determine the absolute configuration of 2,3-epoxy-2-methylbutanoate ester residues in natural products is presented, based on (i) the reduction of the ester function to yield a 2-methyl-1,2-butanediol, (ii) esterification of the obtained primary alcohol with either (R)-(+)- or
Giancarlo Cravotto et al.
Chirality, 16(8), 526-533 (2004-08-04)
Cyclodextrin (CD) derivatives are important selectors for analytical chiral recognition. Their enantioselectivities and chemical properties depend on ring size and on nature, number and location of substituents. This paper describes the synthesis of 6-O-TBDMS-2,3-O-methyl beta-cyclodextrins bearing in position 2 either
Hiroshi Hasegawa et al.
Journal of mass spectrometry : JMS, 46(5), 502-507 (2011-04-19)
D-Serine is a co-agonist of the N-methyl-D-aspartate receptor in glutamate neurotransmission and has been proposed as a potential therapeutic agent for schizophrenia. However, D-serine also acts as a nephrotoxic substance in rats at high doses. To investigate the pharmacokinetics and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service