Skip to Content
Merck
All Photos(1)

Documents

62120

Sigma-Aldrich

(R)-(+)-Limonene

purum, ≥96.0% (sum of enantiomers, GC)

Synonym(s):

(+)-p-Mentha-1,8-diene, (+)-Carvene, (R)-4-Isopropenyl-1-methyl-1-cyclohexene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
2204754
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

4.7 (vs air)

vapor pressure

<3 mmHg ( 14.4 °C)

grade

purum

Assay

≥96.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D +112±3°, c = 10% in ethanol

expl. lim.

6.1 %

refractive index

n20/D 1.473 (lit.)
n20/D 1.473

bp

176-177 °C (lit.)

density

0.842 g/mL at 20 °C (lit.)

SMILES string

CC(=C)[C@@H]1CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3/t10-/m0/s1

InChI key

XMGQYMWWDOXHJM-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cedric S Graebin et al.
European journal of medicinal chemistry, 45(4), 1524-1528 (2010-02-02)
The synthesis and in vitro activity of R(+)-Limonene derivatives against Leishmania and Trypanosoma cruzi strains are reported. Seven compounds have shown better in vitro activity against Leishmania (V.)braziliensis than the standard drug pentamidine. Additionally, we have identified two promising new
Bharat B Aggarwal et al.
Biochemical pharmacology, 71(10), 1397-1421 (2006-03-28)
While fruits and vegetables are recommended for prevention of cancer and other diseases, their active ingredients (at the molecular level) and their mechanisms of action less well understood. Extensive research during the last half century has identified various molecular targets
W G Flamm et al.
Regulatory toxicology and pharmacology : RTP, 13(1), 70-86 (1991-02-01)
The monoterpene d-limonene is a naturally occurring chemical which is the major component in oil of orange. Currently, d-limonene is widely used as a flavor and fragrance and is listed to be generally recognized as safe (GRAS) in food by
Patrizia A d'Alessio et al.
Life sciences, 92(24-26), 1151-1156 (2013-05-15)
To further explore the anti-inflammatory properties of d-Limonene. A rat model was used to compare evolution of TNBS (2,5,6-trinitrobenzene sulfonic acid)-induced colitis after oral feeding with d-Limonene compared to ibuprofen. Peripheral levels of TNF-α (Tumor Necrosis Factor alpha) were assessed
Young Woo Kim et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 16(1), 17-38 (2013-04-12)
d-Limonene, a major constituent of citrus oils, is a monoterpene widely used as a flavor/fragrance additive in cosmetics, foods, and industrial solvents as it possesses a pleasant lemon-like odor. d-Limonene has been designated as a chemical with low toxicity based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service