Skip to Content
Merck
All Photos(1)

Key Documents

61852

Supelco

Vanillyl alcohol

analytical standard

Synonym(s):

4-Hydroxy-3-methoxybenzyl alcohol, NSC 3993, Vanillyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH2OH
CAS Number:
Molecular Weight:
154.16
Beilstein:
1910044
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

110-117 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1cc(CO)ccc1O

InChI

1S/C8H10O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-4,9-10H,5H2,1H3

InChI key

ZENOXNGFMSCLLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vanillyl alcohol belongs to the phenolic group of compounds which are widely used as oxidants in food products.

Application

Vanillyl alcohol may be used as a reference standard for the determination of vanillyl alcohol in:
  • Processed lignin samples by ultra-high performance supercritical fluid chromatography combined with quadrupole time-of-flight mass spectrometry (UHPSFC-QTOF-MS).
  • Herb liqueurs by gas chromatography-flame ionization detection (GC-FID), GC with mass spectrometry operating in electron impact (EI) mode and high performance liquid chromatography-ultraviolet (HPLC-UV) detection.
  • Aqueous extracts of sesame (Sesanum indicum) oil by LC with tandem mass spectrometry (MS/MS) equipped with electrospray ionization source (ESI).
  • Tall gastrodia tuber by HPLC-UV.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of gastrodin, p-hydroxybenzyl alcohol, vanillyl alcohol, p-hydroxylbenzaldehyde and vanillin in tall gastrodia tuber by high-performance liquid chromatography.
Liu CL, et al.
Chromatographia, 55(5-6), 317-320 (2002)
Identification and evaluation of anti-inflammatory properties of aqueous components extracted from sesame (Sesamum indicum) oil.
Deme P, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1087(4), 61-69 (2018)
Enzymatic production of bioactive docosahexaenoic acid phenolic ester.
Roby MH, et al.
Food Chemistry, 171, 397-404 (2015)
Phenolic compounds and aroma-impact odorants in herb liqueurs elaborated by maceration of aromatic and medicinal plants in grape marc distillates.
Rodriguez-Solana R, et al.
Journal of the Institute of Brewing, 122(4), 653-660 (2016)
Ultra-high-performance supercritical fluid chromatography with quadrupole-time-of-flight mass spectrometry (UHPSFC/QTOF-MS) for analysis of lignin-derived monomeric compounds in processed lignin samples.
Prothmann J, et al.
Analytical and Bioanalytical Chemistry, 409(30), 7049-7061 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service