Skip to Content
Merck
All Photos(1)

Key Documents

59962

Supelco

Conductivity Standard

conductivity 5000 μS/cm at 25 °C

Synonym(s):

Calibration standard for conductivity meter

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€161.00

€161.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
€161.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€161.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

≤2.5% KCl (T)

conductivity

4950-5050 μS/cm at 25 °C

density

0.995-1.014 g/cm3 at 20 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

single component solution

Looking for similar products? Visit Product Comparison Guide

Application

Calibration standard for conductivity measuring instruments.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liptak.GB and Venczel K
Analysis and Analyzers, Volume 2 (2016)
Volunteer stream monitoring a methods manual null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service