Skip to Content
Merck
All Photos(1)

Key Documents

58689

Supelco

N-Isobutyryl-D-cysteine

for chiral derivatization, LiChropur, ≥97.0%

Synonym(s):

N-(2-Methylpropionyl)-D-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH[NHCOCH(CH3)2]CO2H
CAS Number:
Molecular Weight:
191.25
MDL number:
UNSPSC Code:
12000000
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

Quality Level

product line

ChiraSelect

Assay

≥97.0% (RT)
≥97.0%

form

solid

optical purity

enantiomeric ratio: ≥99.5:0.5 (HPLC)

quality

LiChropur

mp

97-101 °C (lit.)
97-101 °C

storage temp.

2-8°C

SMILES string

CC(C)C(=O)N[C@H](CS)C(O)=O

InChI

1S/C7H13NO3S/c1-4(2)6(9)8-5(3-12)7(10)11/h4-5,12H,3H2,1-2H3,(H,8,9)(H,10,11)/t5-/m1/s1

InChI key

BWBQXMAXLAHHTK-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

It was used for derivatization of amino acid mixtures of OPA during HPLC analysis of L- and D-amino acids in plants.[1]
N-Isobutyryl-D-cysteine is a chiral thiol mostly used in precolumn orthophthaldehyde (OPA) derivatization of amino acids.[2]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographic determination of L- and D-amino acids in plants.
Bruckner H and Westhauser T.
Amino Acids, 24 (1-2), 43-55 (2003)
Handbook of HPLC
Danilo Corradini
Science, 715-715 (2011)
Hui Gan et al.
Angewandte Chemie (International ed. in English), 48(29), 5282-5286 (2009-03-25)
The right fit: Plasmid DNA molecules show chirality-dependent interaction with gold surfaces modified by L and D N-isobutyrylcysteine. Relaxed DNA molecules have a stronger interaction and adsorption on the L surface, while their counterparts on the D surface maintain a
Stereospecific interaction between immune cells and chiral surfaces.
Taolei Sun et al.
Journal of the American Chemical Society, 129(6), 1496-1497 (2007-02-08)
H Brückner et al.
Journal of chromatography. A, 711(1), 201-215 (1995-09-08)
In order to evaluate and extend the applicability of an analytical method that enables the quantitative and simultaneous high-performance liquid chromatographic determination of D- and L-amino acids (DL-AAs) by automated precolumn derivatization with o-phthaldialdehyde together with the chiral thiol N-isobutyryl-L-cysteine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service