Skip to Content
Merck
All Photos(1)

Key Documents

573701

Supelco

Dowex SBR-C Chloride form

chloride form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000
To inquire about this product 573701, please contact your local Merck office or dealer. Contact Technical Service

grade

condensate grade

Quality Level

description

cross-linkage 8%

form

beads

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

quaternary ammonium

particle size

16-45 mesh

operating pH

0-14

capacity

1.4 meq/mL by wetted bed volume

bulk density

705 g/L (20.0 kg/cuft)

separation technique

anion exchange

Application

Strongly basic (type I) anion exchange resin with a particle size distribution suited to deep-bed condensate polishing and mixed-bed applications. This resin is useful for general demineralization and meets the requirements of FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service