Skip to Content
Merck
All Photos(1)

Key Documents

56332

Sigma-Aldrich

(2-Hydroxypropyl)-β-cyclodextrin

Mr ~1380

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

optical activity

[α]20/D +135±10°, c = 1% in H2O

mol wt

Mr ~1380

extent of labeling

~0.6 substitution

mp

~278 °C (dec.)

SMILES string

CC(O)COCC1OC2OC3C(COCC(C)O)OC(OC4C(COCC(C)O)OC(OC5C(COCC(C)O)OC(OC6C(COCC(C)O)OC(OC7C(COCC(C)O)OC(OC8C(COCC(C)O)OC(OC1C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C8OCC(C)O)C(OCC(C)O)C7OCC(C)O)C(OCC(C)O)C6OCC(C)O)C(OCC(C)O)C5OCC(C)O)C(OCC(C)O)C4OCC(C)O)C(OCC(C)O)C3OCC(C)O

InChI

1S/C63H112O42/c1-22(64)8-85-15-29-50-36(71)43(78)57(92-29)100-51-30(16-86-9-23(2)65)94-59(45(80)38(51)73)102-53-32(18-88-11-25(4)67)96-61(47(82)40(53)75)104-55-34(20-90-13-27(6)69)98-63(49(84)42(55)77)105-56-35(21-91-14-28(7)70)97-62(48(83)41(56)76)103-54-33(19-89-12-26(5)68)95-60(46(81)39(54)74)101-52-31(17-87-10-24(3)66)93-58(99-50)44(79)37(52)72/h22-84H,8-21H2,1-7H3/t22?,23?,24?,25?,26?,27?,28?,29-,30-,31?,32?,33?,34?,35?,36?,37-,38?,39-,40+,41+,42+,43?,44+,45?,46+,47+,48+,49+,50+,51+,52-,53-,54-,55-,56-,57+,58-,59+,60-,61-,62-,63-/m1/s1

InChI key

ODLHGICHYURWBS-RYJYQAAZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The solubility of lipophilic drugs increases linearly with the concentration of hydroxypropyl-β-cyclodextrin (HBC) in aqueous solution because of the complex between HBC and the drug. This guest-host type complex is formed between the drug and the non-polar cavity in the HBC that results in enhanced solubility. Solutions may be lyophilized to produce freely soluble powders. Non-toxic in rabbits and mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Zhong Wang et al.
Mediators of inflammation, 2011, 848309-848309 (2011-03-16)
Our previous study concerning brain trauma has shown that progesterone could regulate toll-like receptor 4 (TLR4) and nuclear factor-kappa B (NF-κB) signaling pathway in the brain, which also has been proved to play important roles in early brain injury (EBI)
Dmitri R Davydov et al.
Biochemistry, 47(43), 11348-11359 (2008-10-04)
To establish a direct method for monitoring substrate binding in cytochrome P450eryF applicable at elevated hydrostatic pressures, we introduce a laser dye Fluorol-7GA (F7GA) as a novel fluorescent ligand. The high intensity of fluorescence and the reasonable resolution of the
Shuhua Chen et al.
PloS one, 6(8), e24293-e24293 (2011-09-16)
Previously, we demonstrated that allopregnanolone (APα) promoted proliferation of rodent and human neural progenitor cells in vitro. Further, we demonstrated that APα promoted neurogenesis in the hippocampal subgranular zone (SGZ) and reversed learning and memory deficits in the male triple
Rachel J Smith et al.
The European journal of neuroscience, 30(3), 493-503 (2009-08-07)
The orexin/hypocretin system has recently been implicated in reward-processing and addiction. We examined the involvement of the orexin system in cue-induced reinstatement of extinguished cocaine-seeking by administering the orexin 1 receptor antagonist SB-334867 (SB) or the orexin 2 receptor antagonist
D E Rusyniak et al.
Neuroscience, 154(4), 1619-1626 (2008-06-07)
When given in a warm environment MDMA (3,4-methylenedioxymethamphetamine, ecstasy) causes hyperthermia by increasing interscapular brown adipose tissue (iBAT) heat production and decreasing heat loss via cutaneous vasoconstriction. When given in a cold environment, however, MDMA causes hypothermia by an unknown

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service