Skip to Content
Merck
All Photos(1)

Key Documents

544167

Sigma-Aldrich

Cobalt(II) sulfate heptahydrate

technical grade

Synonym(s):

Cobaltous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoSO4 · 7H2O
CAS Number:
Molecular Weight:
281.10
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

technical grade

density

2.03 g/mL at 25 °C (lit.)

SMILES string

O.[Co++].[O-]S([O-])(=O)=O

InChI

1S/Co.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

BGORGFZEVHFAQU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cobalt sulfate.
Report on carcinogens : carcinogen profiles, 12, 113-115 (2011-08-19)
Carolyn F Moyer et al.
Toxicologic pathology, 30(4), 427-434 (2002-08-22)
Epidemiological studies suggest an association between ambient particulate matter and cardiopulmonary diseases in humans. The mechanisms underlying these health effects are poorly understood. To better understand the potential relationship between particulate-matter-induced inflammation and vascular disease, a 2-phase retrospective study was
Environment toxicity as a new universal characteristic of the pollution of objects tested.
P E Tulupov et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 399, 496-499 (2005-02-19)
S Sahoo et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 26(12), 1309-1312 (2010-12-16)
An analytical method has been described for the determination of mercury using a carbon paste electrode. Conditions for the preparation of the carbon paste electrode were optimized for low background current in order to use it for the determination of
Lei Hou et al.
Chemical communications (Cambridge, England), 46(34), 6311-6313 (2010-08-03)
Solvothermal reaction of CoSO(4) with 2,6-di-p-carboxyphenyl-4,4'-bipyridine affords a novel, octacobalt cluster based, (3,12)-connected porous framework, which exhibits gas sorption and spin-glassy magnetic behaviour.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service