Skip to Content
Merck
All Photos(1)

Key Documents

537543

Sigma-Aldrich

Propylene glycol monomethyl ether acetate

99%

Synonym(s):

1,2-Propanediol monomethyl ether acetate, 1-Methoxy-2-propyl acetate, MPA, PGMEA, Propylene glycol methyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH(CH3)CH2OCH3
CAS Number:
Molecular Weight:
132.16
Beilstein:
1751656
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

3.7 mmHg ( 20 °C)

Assay

99%

autoignition temp.

669 °F

expl. lim.

13.1 %

refractive index

n20/D 1.402 (lit.)

bp

145-146 °C (lit.)

density

0.970 g/mL at 25 °C (lit.)

SMILES string

COCC(C)OC(C)=O

InChI

1S/C6H12O3/c1-5(4-8-3)9-6(2)7/h5H,4H2,1-3H3

InChI key

LLHKCFNBLRBOGN-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

replaced by

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

113.9 °F - closed cup

Flash Point(C)

45.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Limor Cohen et al.
ACS nano, 14(8), 9491-9501 (2020-06-27)
Many proteins are present at low concentrations in biological samples, and therefore, techniques for ultrasensitive protein detection are necessary. To overcome challenges with sensitivity, the digital enzyme-linked immunosorbent assay (ELISA) was developed, which is 1000× more sensitive than conventional ELISA
Wushuang Wan et al.
Nature communications, 11(1), 4199-4199 (2020-08-23)
Aqueous-phase approaches to semiconductor CdS magic-size clusters (MSCs) and the formation pathway have remained relatively unexplored. Here, we report the demonstration of an aqueous-phase, room-temperature approach to CdS MSCs, together with an exploration of their evolution pathway. The resulting CdS
Charles A Conn et al.
Lab on a chip, 14(20), 3968-3977 (2014-08-13)
Foam mobility control and novel oil displacement mechanisms were observed in a microfluidic device representing a porous media system with layered permeability. Foam was pre-generated using a flow-focusing microfluidic device and injected into an oil-wet, oil-saturated 2-D PDMS microfluidic device.
Simon Braun et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(12), 3641-3646 (2015-03-17)
The dynamics of quantum phase transitions pose one of the most challenging problems in modern many-body physics. Here, we study a prototypical example in a clean and well-controlled ultracold atom setup by observing the emergence of coherence when crossing the
Shaohua Ma et al.
Lab on a chip, 14(18), 3611-3620 (2014-07-30)
The flow topology in moving microdroplets has a significant impact on the behaviour of encapsulated objects and hence on applications of the technology. This study reports on a systematic investigation of the flow field inside droplets moving in a rectangular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service