Skip to Content
Merck
All Photos(1)

Key Documents

53024

Supelco

Lauryl gallate

analytical standard

Synonym(s):

E312, Dodecyl gallate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O5
CAS Number:
Molecular Weight:
338.44
Beilstein:
2701981
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E312
NACRES:
NA.24

Quality Level

grade

analytical standard

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

94-97 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C19H30O5/c1-2-3-4-5-6-7-8-9-10-11-12-24-19(23)15-13-16(20)18(22)17(21)14-15/h13-14,20-22H,2-12H2,1H3

InChI key

RPWFJAMTCNSJKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lauryl gallate is a major monomer flavanol found in green tea and shows antioxidant activity in polymers.[1]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Lauryl gallate may be used as an analytical standard in investigating its activity as a pro-ecological anti-ageing substance for common polymers as food packing materials using electroanalytical techniques such as cyclic voltammetry (CV) and differential pulse voltammetry (DPV) methods.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dodecyl gallate as a pro-ecological antioxidant for food packing materials
Masek A, et al.
Comptes Rendus Chimie, 17(11), 1116-1127 (2014)
Jordi Garcia-Ubasart et al.
Bioresource technology, 112, 341-344 (2012-03-24)
A new biotechnological procedure using laccase in combination with a hydrophobic phenolic compound (lauryl gallate) for the hydrophobization of cellulose fibres and internal sizing of paper was developed. Cellulose fibres from hardwood kraft pulp were incubated with laccase (Lac), in
K Fujita et al.
Journal of applied microbiology, 92(6), 1035-1042 (2002-05-16)
The aim was to investigate the antifungal actions of nonyl gallate against Saccharomyces cerevisiae ATCC 7754. The maximum potency of both the growth inhibitory and the fungicidal effect against the yeast strain was found in nonyl gallate among n-alkyl gallates
Contact allergy to propylene glycol and dodecyl gallate mimicking seborrheic dermatitis.
Alexander V Kuznetsov et al.
Contact dermatitis, 55(5), 307-308 (2006-10-10)
Carolina Hurtado et al.
Antiviral therapy, 13(7), 909-917 (2008-12-03)
Antiviral compounds are needed in the control of many animal and human diseases. We analysed the effect of the antitumoural drug lauryl gallate on the infectivity of the African swine fever virus among other DNA (herpes simplex and vaccinia) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service