Skip to Content
Merck
All Photos(1)

Key Documents

52908

Supelco

Hexapentacontane

purum p.a., standard for GC, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)54CH3
CAS Number:
Molecular Weight:
787.50
Beilstein:
5142319
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

standard for GC
purum p.a.

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

96-99 °C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service