Skip to Content
Merck
All Photos(1)

Key Documents

52451

Supelco

Sodium benzoate

Sodium benzoate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Benzoic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€85.50

€85.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€85.50

About This Item

Linear Formula:
C6H5COONa
CAS Number:
Molecular Weight:
144.10
Beilstein:
3572467
EC Number:
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.24

€85.50


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

WXMKPNITSTVMEF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application


  • Sodium benzoate preservative efficacy research: A study employing dicationic imidazole ionic liquid stationary phases demonstrated enhanced detection capabilities for preservatives including sodium benzoate, utilizing a mixed mode of Hydrophilic Interaction Liquid Chromatography (HILIC), Reversed-Phase Liquid Chromatography (RPLC), and Ion Exchange Chromatography (IEC). This application is critical for maintaining food safety and quality in the food industry (Yang et al., 2024).

  • Sodium benzoate metabolic pathway studies: The metabolic pathways of sodium benzoate were explored in relation to its efficacy and safety in food and pharmaceutical products. These studies are pivotal for understanding the metabolic breakdown and potential toxicity of sodium benzoate, ensuring compliance with health regulations and consumer safety (Yang et al., 2024).

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service